Funnels with short stem, PYREX®
供应商: CORNING SCIENCE PRODUCTS
6180-65 6180-65_U
CORN6180-65EA 0 CNY
CORN6180-65 CORN6180-65_U
Funnels with short stem, PYREX®
漏斗 过滤漏斗 Filter Funnels
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With a beaded edge for strength. Cone angle 60°.