Paraffin Oil, J.T.Baker®
供应商: Avantor
BAKRS894-00EA 2481 CNY
BAKRS894-00 BAKRS894-07
Paraffin Oil, J.T.Baker®
Paraffin Oil
沸点: 300…500 °C (1013 hPa)
密度: 0,86 g/cm³ (20 °C)
闪点: 215 °C (closed cup)
CAS号: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07093
现在订购
产品技术规格
For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test
Product Information (not specifications): Saybolt Viscosity @ 100
degrees F (sec).... 345 - 355