Volumetric pipettes, color-coded, class A, PYREX®
供应商: CORNING SCIENCE PRODUCTS
7100-1 7100-10 7100-2 7100-25 7100-4 7100-50
CORN7100-1EA 0 CNY
CORN7100-1 CORN7100-10 CORN7100-2 CORN7100-25 CORN7100-4 CORN7100-50
Volumetric pipettes, color-coded, class A, PYREX®
移液管和移液器 刻度移液管
Colored markings remain sharply legible for the life of pipette.
  • Reusable, rugged construction
  • Colour-coded
  • Coloured enamel graduations and markings
Ruggedly constructed and properly proportioned, pipettes have accuracy tolerances to meet or exceed the strict requirements for Class A Volumetric Ware as indicated by ASTM E969.