2,6-Dichlorophenolindophenol sodium salt, powder, AR® ACS, Macron Fine Chemicals™

供应商: Avantor Performance Materials

Synonyms: Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide, Tillmans' reagent, 2,6-Dichloroindophenol sodium, 2,6-Dichloroindophenol sodium salt, 2,6-Dichloroindophenol sodium derivative

MACR3054-54EA 0 CNY
MACR3054-54
2,6-Dichlorophenolindophenol sodium salt, powder, AR® ACS, Macron Fine Chemicals™
2,6-Dichlorophenolindophenol sodium salt

Meets ACS Reagent Chemical Requirements. For Laboratory, Research, or Manufacturing Use

分子式: C₁₂H₆Cl₂NNaO₂
分子量: 290.08 g/mol
存储条件: Ambient
CAS号: 620-45-1
EINECS: 210-640-4
Merck Index: 13,03093

现在订购

产品技术规格

Meets ACS Reagent Chemical Requirements = 12.0 %
For Laboratory,Research,or Manufacturing Use Passes Test
ACS - Loss on Drying at 120°C ≤12.0 %
ACS - Interfering Dyes Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR