Volumetric flasks, class A, low actinic, glass stopper, PYREX®
供应商: CORNING SCIENCE PRODUCTS
55680-10 55680-100 55680-25 55680-50
CORN55680-10EA 0 CNY
CORN55680-10 CORN55680-100 CORN55680-25 CORN55680-50
Volumetric flasks, class A, low actinic, glass stopper, PYREX®
烧瓶 容量瓶
PYREX®, borosilicate glass, amber, low actinic, with tapered ground stopper.
  • Individually serialised
  • Markings are sharp and permanent
  • Low actinic glass provide protection for material sensitive to light
Each flask is individually serialised and supplied with a Certificate of Identification and Capacity, traceable to NIST standards. The protective color is an integral part of the flask, which retains its mechanical strength, chemical stability and thermal resistance.
Certifications: Calibrated to meet ASTM specifications E-694, E-542, and E-288