VWR®, 冰盘,有盖或无盖
供应商: VWR
VWR®
VWRI216-1533EA 0 CNY
VWRI216-1533 VWRI216-1532 VWRI216-1499 VWRI216-1531 VWRI216-1498 VWRI216-1527 VWRI216-1538 VWRI216-1526 VWRI216-1537 VWRI216-1525 VWRI216-1503 VWRI216-1509 VWRI216-1508 VWRI216-1529 VWRI216-1507 VWRI216-1528 VWRI216-1506 VWRI216-1539 VWRI216-1493 VWRI216-1497 VWRI216-1530 VWRI216-1496 VWRI216-1495 VWRI216-1494
VWR®, 冰盘,有盖或无盖
储冰容器
Polyurethane foam.
  • Resistant to temperatures from –20 to +70 °C
  • Extremely robust, leakproof, stackable and lightweight
  • Available in three sizes and four colours
  • Stacking option for space saving on working bench
  • Pouring lip design, for easy draining of liquid
  • Lid to reduce the evaporation level
  • 可耐受–20到+70 °C温度
  • 极其结实,防漏,可堆叠,轻量
  • 三个尺寸和四种颜色可选
  • 可堆叠,节省工作台空间
  • 浇注嘴设计,便于排出液体
  • 加盖设计,减少带盐泥的干冰和水冰的蒸发水平
Ideal for storing samples, these pans maintain hot or cold temperatures of bottles, test tubes, solutions and cultures. Suitable for use with dry ice, water ice and salt solution. For freezer storage only.