4-氨基-3-羟基-1-萘磺酸, OR, Macron Fine Chemicals™
供应商: Avantor Performance Materials
Synonyms:
1-氨基-2-萘酚-4-磺酸
MACR1807-55EA
0
CNY
MACR1807-55
4-氨基-3-羟基-1-萘磺酸, OR, Macron Fine Chemicals™
4-氨基-3-羟基-1-萘磺酸
Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications.
分子式:
C₁₀H₉NO₄S 分子量: 239.25 g/mol 沸点: 466 溶点: ∼285 °C 存储条件: Ambient |
MDL编号:
MFCD00004019 CAS号: 116-63-2 EINECS: 204-147-3 Merck Index: 13,00453 |
产品技术规格
For Laboratory,Research,or Manufacturing Use | |
Identification | Passes Test |
Appearance (purple powder) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...