Chemical resistant gloves, AlphaTec® 58-430 / 58-435
供应商: Ansell
58-435/10 58-435/7 58-435/11 58-435/8 58-435/9 58-430/10 58-430/11 58-430/7 58-430/8 58-430/9
ANSE58-435/10EA 0 CNY
ANSE58-435/10 ANSE58-435/7 ANSE58-435/11 ANSE58-435/8 ANSE58-435/9 ANSE58-430/10 ANSE58-430/11 ANSE58-430/7 ANSE58-430/8 ANSE58-430/9
Chemical resistant gloves, AlphaTec® 58-430 / 58-435
手套
These gauntlet style gloves feature Ansell Grip Technology™ for optimal control when handling oily or wet items. Less force is required on slippery objects, reducing hand and arm fatigue.
  • Medium duty 58-430 features a 10 mil (~0,25 mm) shell to protect against exposure to bases, oils, fuels, some solvents, greases and animal fats
  • Heavy duty 58-435 has a 16 mil (~0,41 mm) shell effective against hydrocarbon derivatives, alcohol bases, many solvents and esters
  • Nitrile coating offers superior snag, puncture and abrasion protection for long-term wear
  • Foldable gutter cuff helps prevent dripping onto the forearm
  • Extra-long cuff (58-435) extends protection to the upper forearm
Both models offer improved flexibility and dexterity thanks to a cotton flocked-lined nitrile shell and snug, second-skin tailoring that tightens the fit across the palm.
Colour: Green/grey
58-430: EN 374 (JKL), EN 388 (3001), EN 1149; Cat. III; AQL 0,65 58-435: EN 374 (AJKL), EN 388 (4001), EN 1149; Cat. III; AQL 0,65 Food contact approved
Certifications: 58-430: EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN 388:2016 (3001X), EN 1149; Cat. III; AQL 0,65
58-435: EN ISO 374-1:2016 Type A (AJKLOPT), EN ISO 374-5:2016, EN 388:2016 (4001A), EN 1149; Cat. III; AQL 0,65
Food contact approved