Industrial gloves, AlphaTec® Solvex® 37-175
供应商: Ansell
37-175-10 37-175-7 37-175-9
ANSE37-175-10EA 0 CNY
ANSE37-175-10 ANSE37-175-7 ANSE37-175-9
Industrial gloves, AlphaTec® Solvex® 37-175
手套
These flock-lined gloves made of a high performance nitrile compound provide an outstanding combination of chemical resistance and mechanical strength.
  • Offers superior snag, puncture and abrasion protection over rubber or neoprene gloves
  • Suitable for heavy-duty cleaning applications
  • High levels of flexibility, comfort and dexterity
  • Won't swell, weaken or degrade, and does not promote contact dermatitis
  • Straight cuff, sandpatch finish
Suitable for heavy-duty cleaning applications, food processing, janitorial and maintenance work.
Thickness: 0,4 mm
Colour: Green
EN 388 (3101) Food contact approved
Certifications: EN 388:2016 (4101X), EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5 VIRUS; Cat. III
Food contact approved