Polyadenylic acid (poly-A)
供应商: Cytiva
27-4110-01 27-4110-02
GEHE27-4110-01EA 0 CNY
GEHE27-4110-01 GEHE27-4110-02
Polyadenylic acid (poly-A)
载体
Poly(A) (polyadenylic acid) can be used to evaluate the binding of potential therapeutic agents to mRNA in drug development research.
  • Average chain length of >200 nucleotides
  • Ribonuclease tested