J.T.Baker® BAKERBOND Speedisk®, Extraction Discs
供应商: Avantor
BAKR8055-06EA 5516 CNY
BAKR8055-06 BAKR8055-07 BAKR8056-06 BAKR8057-06 BAKR8058-06 BAKR8060-06 BAKR8061-06 BAKR8068-06 BAKR8068-07 BAKR8072-06 BAKR8072-07 BAKR8082-06
J.T.Baker® BAKERBOND Speedisk®, Extraction Discs
过滤器 过滤盘
Discs are pre-assembled for use in preparing aqueous samples for analysis. Laminar configuration provides filtration capacity and inlet characteristics that maximise access of analyte molecules to the micro-particulate sorbent. Disc design resists clogging and ensures high throughput rates even when samples contain solids.