J.T.Baker® BAKERBOND spe™ SiOH, SPE Columns
供应商: Avantor
BAKR7086-03EA 1858 CNY
BAKR7086-03 BAKR7086-01 BAKR7086-06 BAKR7086-23 BAKR7086-26 BAKR7086-07
J.T.Baker® BAKERBOND spe™ SiOH, SPE Columns
色谱柱
BAKERBOND spe™ columns are disposable PP columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
General applications: Polar analytes from non polar solvents like hydrocarbons and less polar esters and ethers.
These SPE columns can be used in adsorption mode.
Packaging: Certain columns are available in a foil-lined fibre drum called the Jumbo Pack. The columns are loose packed in the foil liner to eliminate the inconvenience of opening individual pouches.