Dry vacuum and chemical resistant diaphragm pumps
供应商: GARDNER DENVER
4000511-04 4000481-04
ILMV4000511-04EA 0 CNY
ILMV4000511-04 ILMV4000481-04
Dry vacuum and chemical resistant diaphragm pumps
真空隔膜泵
These are certified pumps for the evacuation of combustible atmospheres inside or outside of highly combustible environments. They are used for operating in potentially explosive atmospheres.
Certifications: Complies to 1999/92/EG (ATEX 137).