Dissection boards
供应商: BOCHEM
12616 12615
BOCH12616EA 0 CNY
BOCH12616 BOCH12615
Dissection boards
解剖板
Hygienic dissection boards made from PE.
  • Two colours available
  • Suitable for dish washer - Heat resistant up to 90 °C
HACCP compliant
Certifications: HACCP compliant