VWR® ATC, Handheld Refractometers
供应商: VWR Collection
VWRI635-0629EA 1042 CNY
VWRI635-0629 VWRI635-0630 VWRI635-0631 VWRI635-0632 VWRI635-0633 VWRI635-0634 VWRI635-0635 VWRI635-0636 VWRI635-0637 VWRI635-0638 VWRI635-0639
VWR® ATC, Handheld Refractometers
折光仪 手持式折光仪
该款带自动温度补偿(ATC)的手持式折射计能够显示精确而一致的结果。
  • ATC at 20 °C between 10 and 30 °C
  • Scales are easy to read and deliver exact results
  • Easy to use and clean