4-(2-(Dimethylamino)ethyl)phenol ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
Hordenine
H1351-5G
H1351-1G
TCIAH1351-5GEA
0
CNY
TCIAH1351-5G
TCIAH1351-1G
4-(2-(Dimethylamino)ethyl)phenol ≥98.0% (by GC, titration analysis)
4-(2-(Dimethylamino)ethyl)phenol
分子式:
C₁₀H₁₅NO 沸点: 174°C |
MDL编号:
MFCD00051462 CAS号: 539-15-1 EINECS: 208-710-4 Merck Index: 14,04746 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...