Fenclorim ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4,6-Dichloro-2-phenylpyrimidine
F0851-5G
F0851-1G
TCIAF0851-5GEA
0
CNY
TCIAF0851-5G
TCIAF0851-1G
Fenclorim ≥98.0% (by GC)
Fenclorim
分子式:
C₁₀H₆Cl₂N₂ |
MDL编号:
MFCD00754193 CAS号: 3740-92-9 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...