3-三氟甲基吡啶 ≥98.0% (通过GC)

供应商: TCI
Warning

T2786-5G T2786-25G
TCIAT2786-5GEA 0 CNY
TCIAT2786-5G TCIAT2786-25G
3-三氟甲基吡啶 ≥98.0% (通过GC)
3-三氟甲基吡啶
分子式: C₆H₄F₃N
沸点: 113°C
闪点: 23°C
MDL编号: MFCD00100032
CAS号: 3796-23-4
EINECS: 223-268-2
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR