正十二腈 ≥98.0% (通过GC)
供应商: TCI
|
Warning
|
L0110-25ML
L0110-500ML
TCIAL0110-25MLEA
0
CNY
TCIAL0110-25ML
TCIAL0110-500ML
正十二腈 ≥98.0% (通过GC)
正十二腈
分子式:
C₁₂H₂₃N 沸点: 276°C 闪点: 114°C |
MDL编号:
MFCD00001946 CAS号: 2437-25-4 EINECS: 219-440-1 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...