Ethylboronic acid (contains varying amounts of Anhydride)

供应商: TCI
Warning

E0913-5G E0913-1G
TCIAE0913-5GEA 0 CNY
TCIAE0913-5G TCIAE0913-1G
Ethylboronic acid (contains varying amounts of Anhydride)
Ethylboronic acid
分子式: C₂H₇BO₂
MDL编号: MFCD01074536
CAS号: 4433-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=028875f4bfcecc1e79058e9d23cc46916c4e8590&vid=028875f4bfcecc1e79058e9d23cc46916c4e8590&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 133.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR