4-氟-3-(三氟甲基)苯甲酰氯 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

F0659-1G F0659-5G
TCIAF0659-1GEA 0 CNY
TCIAF0659-1G TCIAF0659-5G
4-氟-3-(三氟甲基)苯甲酰氯 ≥98.0% (by GC, titration analysis)
4-氟-3-(三氟甲基)苯甲酰氯
分子式: C₈H₃ClF₄O
分子量: 226.56 g/mol
沸点: 90°C
密度: 1.493
闪点: 77°C
MDL编号: MFCD00061159
CAS号: 67515-56-4
UN号: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e95fec51fa8b1e160d0a8139fdb809f9def2ac1&vid=2e95fec51fa8b1e160d0a8139fdb809f9def2ac1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR