3-氟-5-(三氟甲基)苯甲酰氯 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

F0767-1G F0767-5G
TCIAF0767-1GEA 0 CNY
TCIAF0767-1G TCIAF0767-5G
3-氟-5-(三氟甲基)苯甲酰氯 ≥98.0% (by GC, titration analysis)
3-氟-5-(三氟甲基)苯甲酰氯
分子式: C₈H₃ClF₄O
分子量: 226.56 g/mol
沸点: 119
密度: 1.482
闪点: 74°C
MDL编号: MFCD00061157
CAS号: 171243-30-4
UN号: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27e4aaf5883cb717fc6a04ae25f884a8fa4bc835&vid=27e4aaf5883cb717fc6a04ae25f884a8fa4bc835&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light red to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR