2-氟-6-(三氟甲基)苯甲酰氯 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 2-Fluoro-6-(trifluoromethyl)benzene-1-carbonyl chloride, 2-Fluoro-6-(trifluoromethyl)benzoylchloride

F0765-5G F0765-1G
TCIAF0765-5GEA 0 CNY
TCIAF0765-5G TCIAF0765-1G
2-氟-6-(三氟甲基)苯甲酰氯 ≥98.0% (by GC, titration analysis)
2-氟-6-(三氟甲基)苯甲酰氯
分子式: C₈H₃ClF₄O
分子量: 226.56 g/mol
密度: 1.465
闪点: 82°C
MDL编号: MFCD00061160
CAS号: 109227-12-5
UN号: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c117bed4a59af2a1d3cfae78f78d3031e765985a&vid=c117bed4a59af2a1d3cfae78f78d3031e765985a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR