2,4,6-三氟苯硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: (2,4,6-Trifluorophenyl)metaboric acid, 2,4,6-Trifluorobenzeneboronic acid, Dihydroxy(2,4,6-trifluorophenyl)borane

T2804-5G T2804-1G
TCIAT2804-5GEA 0 CNY
TCIAT2804-5G TCIAT2804-1G
2,4,6-三氟苯硼酸 (contains varying amounts of Anhydride)
2,4,6-三氟苯硼酸
分子式: C₆H₄BF₃O₂
分子量: 175.9 g/mol
MDL编号: MFCD01863169
CAS号: 182482-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=128a7f0cd4d71a34bb5bd24a54155ab1cdb17c70&vid=128a7f0cd4d71a34bb5bd24a54155ab1cdb17c70&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light red powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR