2,5-二甲基吡咯 ≥98.0% (通过GC)
供应商: TCI
|
Danger
|
D0260-25ML
D0260-5ML
TCIAD0260-25MLEA
0
CNY
TCIAD0260-25ML
TCIAD0260-5ML
2,5-二甲基吡咯 ≥98.0% (通过GC)
2,5-二甲基吡咯
分子式:
C₆H₉N 沸点: 169°C 闪点: 54°C 存储条件: Refrigerator |
MDL编号:
MFCD00005223 CAS号: 625-84-3 EINECS: 210-913-8 UN号: 2929 ADR: 6.1,II |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...