4-Nitropicolinic acid ≥98.0% (通过GC,滴定分析)

供应商: TCI
Warning

N0589-1G N0589-100MG
TCIAN0589-1GEA 0 CNY
TCIAN0589-1G TCIAN0589-100MG
4-Nitropicolinic acid ≥98.0% (通过GC,滴定分析)
4-Nitropicolinic acid
分子式: C₆H₄N₂O₄
MDL编号: MFCD00191562
CAS号: 13509-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1abc6d5ad402985a55f8cc4b8997e2bff9ce19ab&vid=1abc6d5ad402985a55f8cc4b8997e2bff9ce19ab&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR