1-Bromo-2-isopropoxybenzene ≥97.0% (by GC)
供应商: TCI
Synonyms:
2-(2'-Bromophenoxy)propane
B3100-25G
B3100-5G
TCIAB3100-25GEA
0
CNY
TCIAB3100-25G
TCIAB3100-5G
1-Bromo-2-isopropoxybenzene ≥97.0% (by GC)
1-Bromo-2-isopropoxybenzene
分子式:
C₉H₁₁BrO 分子量: 215.09 g/mol 沸点: 73°C |
MDL编号:
MFCD00070759 CAS号: 701-07-5 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...