全氟十二烷酸 ≥92.0% (by GC)

供应商: TCI
Danger

Synonyms: 全氟十二酸, Perfluorododecanoic acid

T2492-1G T2492-5G
TCIAT2492-1GEA 0 CNY
TCIAT2492-1G TCIAT2492-5G
全氟十二烷酸 ≥92.0% (by GC)
全氟十二烷酸
分子式: C₁₂HF₂₃O₂
分子量: 614.10 g/mol
沸点: 245°C
MDL编号: MFCD00198081
CAS号: 307-55-1
EINECS: 206-203-2
UN号: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a61535c517bc7ef5689e3de7beecc2e6fd3f5e0&vid=0a61535c517bc7ef5689e3de7beecc2e6fd3f5e0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 92.0 %
Melting point 107.0 to 113.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR