1,3-联苯氧基苯 ≥98.0% (by GC)
供应商: TCI
Synonyms:
间苯二酚二苯醚
D2662-25G
D2662-5G
TCIAD2662-25GEA
0
CNY
TCIAD2662-25G
TCIAD2662-5G
1,3-联苯氧基苯 ≥98.0% (by GC)
1,3-联苯氧基苯
分子式:
C₁₈H₁₄O₂ 沸点: 189°C |
MDL编号:
MFCD00003036 CAS号: 3379-38-2 EINECS: 222-181-7 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 58.0 to 61.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...