1,4-苯并二噁烷-2-羧酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine-2-carboxylic acid

B3406-25G B3406-5G
TCIAB3406-25GEA 0 CNY
TCIAB3406-25G TCIAB3406-5G
1,4-苯并二噁烷-2-羧酸 ≥98.0% (by GC, titration analysis)
1,4-苯并二噁烷-2-羧酸
分子式: C₉H₈O₄
分子量: 180.16 g/mol
MDL编号: MFCD00084888
CAS号: 3663-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8aa515f3852b2546cc8cd6f871c4bec1b90cf7ee&vid=8aa515f3852b2546cc8cd6f871c4bec1b90cf7ee&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR