1-(4-Chlorophenyl)cyclohexanecarboxylic acid 95%

供应商: Thermo Fisher Scientific

Synonyms: 1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid, 1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid

170760050 170760250
ACRO170760050EA 0 CNY
ACRO170760050 ACRO170760250
1-(4-Chlorophenyl)cyclohexanecarboxylic acid 95%
1-(4-Chlorophenyl)cyclohexanecarboxylic acid
分子式: C₁₃H₁₅ClO₂
溶点: 150…156 °C
存储条件: Ambient
MDL编号: MFCD00019350
CAS号: 58880-37-8
EINECS: 261-481-2

现在订购

产品技术规格

Appearance Light grey Powder and/or granules
Infrared spectrum Conforms
Melting point 150 to 156 °C
Titration with TBAH ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR