4-(4-吗啉)苯甲醛 ≥98.0% (by GC)

供应商: TCI

Synonyms: N-(4-甲酰基苯基)吗啉, 4-吗啉苯甲醛

M1625-1G M1625-25G M1625-5G
TCIAM1625-1GEA 0 CNY
TCIAM1625-1G TCIAM1625-25G TCIAM1625-5G
4-(4-吗啉)苯甲醛 ≥98.0% (by GC)
4-(4-吗啉)苯甲醛
分子式: C₁₁H₁₃NO₂
MDL编号: MFCD00735826
CAS号: 1204-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=991d91963e2a5a3b91f01e6d8ed083b8ab1b7da4&vid=991d91963e2a5a3b91f01e6d8ed083b8ab1b7da4&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 68.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR