Bis(trimethylsilyl)sulphate ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
Bis(trimethylsilyl) sulphate
B1245-25G
B1245-5G
TCIAB1245-25GEA
0
CNY
TCIAB1245-25G
TCIAB1245-5G
Bis(trimethylsilyl)sulphate ≥98.0% (by titrimetric analysis)
Bis(trimethylsilyl)sulphate
分子式:
C₆H₁₈O₄SSi₂ 分子量: 242.4407 g/mol 沸点: 90°C 存储条件: Refrigerator |
MDL编号:
MFCD00008268 CAS号: 18306-29-1 EINECS: 242-192-0 UN号: 2925 ADR: 4.1 III |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...