1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene hydrate ≥98.0% (by GC)
供应商: TCI
|
Warning
|
B1419-25G
B1419-5G
TCIAB1419-25GEA
0
CNY
TCIAB1419-25G
TCIAB1419-5G
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene hydrate ≥98.0% (by GC)
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene hydrate
分子式:
C₁₂H₆F₁₂O₂·H₂O |
MDL编号:
MFCD00192487 CAS号: 1992-15-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 92.0 to 96.0 deg-C |
Water | 3.5 to 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...