2,4,6-Tri-tert-butylnitrosobenzene ≥98.0% (by GC)

供应商: TCI

T1534-1G T1534-100MG
TCIAT1534-1GEA 0 CNY
TCIAT1534-1G TCIAT1534-100MG
2,4,6-Tri-tert-butylnitrosobenzene ≥98.0% (by GC)
2,4,6-Tri-tert-butylnitrosobenzene
分子式: C₁₈H₂₉NO
溶点: 168°C
存储条件: Refrigerator
MDL编号: MFCD00008817
CAS号: 24973-59-9
EINECS: 246-553-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc2f91253e01d047c6cdc01a682547f1bf279f5e&vid=cc2f91253e01d047c6cdc01a682547f1bf279f5e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Green to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR