Methyl 4-methylsalicylate ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Methyl 2-hydroxy-4-methylbenzoate

M1584-25G M1584-5G
TCIAM1584-25GEA 0 CNY
TCIAM1584-25G TCIAM1584-5G
Methyl 4-methylsalicylate ≥98.0% (by GC)
Methyl-4-methylsalicylate
分子式: C₉H₁₀O₃
分子量: 166.18 g/mol
沸点: 242°C
MDL编号: MFCD00020130
CAS号: 4670-56-8
EINECS: 225-117-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=104f6e7bcef3f19f80f4e268a64955ff2b4742ca&vid=104f6e7bcef3f19f80f4e268a64955ff2b4742ca&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Freezing point 24.0 to 28.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR