N-Boc-D-苯丙氨醇 ≥97.0% (by GC)

供应商: TCI

Synonyms: N-(叔丁氧基羰基)-D-苯丙氨醇, (R)-(+)-2-(叔丁氧羰基氨基)-3-苯基-1-丙醇, (R)-2-(Boc-氨基)-3-苯基-1-丙醇

B1966-1G B1966-5G
TCIAB1966-1GEA 0 CNY
TCIAB1966-1G TCIAB1966-5G
N-Boc-D-苯丙氨醇 ≥97.0% (by GC)
N-Boc-D-苯丙氨醇
分子式: C₁₄H₂₁NO₃
分子量: 251.3 g/mol
MDL编号: MFCD00216472
CAS号: 106454-69-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50f4d8c1aebd45f7a3b5ec834d9c073aff17b86e&vid=50f4d8c1aebd45f7a3b5ec834d9c073aff17b86e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Optical purity (LC) min. 98.0 ee%
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR