6-(氯甲基)尿嘧啶 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 6-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione

C1057-25G C1057-5G
TCIAC1057-25GEA 0 CNY
TCIAC1057-25G TCIAC1057-5G
6-(氯甲基)尿嘧啶 ≥98.0% (by HPLC, titration analysis)
6-(氯甲基)尿嘧啶
分子式: C₅H₅ClN₂O₂
分子量: 160.56 g/mol
MDL编号: MFCD00010095
CAS号: 18592-13-7
EINECS: 242-431-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea24c5b1336f2c64c361793d4e276cda8f154afe&vid=ea24c5b1336f2c64c361793d4e276cda8f154afe&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in N,N-DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR