Z-D-Lys-OH ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
Nα-Z-D-赖氨酸, Nα-Cbz-D-赖氨酸
C2136-1G
C2136-5G
TCIAC2136-1GEA
0
CNY
TCIAC2136-1G
TCIAC2136-5G
Z-D-Lys-OH ≥98.0% (by HPLC, titration analysis)
Z-D-Lys-OH
分子式:
C₁₄H₂₀N₂O₄ 分子量: 280.3 g/mol |
MDL编号:
MFCD00067713 CAS号: 70671-54-4 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 215.0 to 219.0 deg-C |
Specific rotation [a]20/D | +11.0 to +15.0 (C=2, 0.2mol/L HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...