4-哌啶甲酰胺 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 异哌啶基甲酰胺, 哌啶-4-甲酰胺

I0255-25G I0255-250G
TCIAI0255-25GEA 0 CNY
TCIAI0255-25G TCIAI0255-250G
4-哌啶甲酰胺 ≥98.0% (by GC)
4-哌啶甲酰胺
分子式: C₆H₁₂N₂O
分子量: 128.18 g/mol
MDL编号: MFCD00038012
CAS号: 39546-32-2
EINECS: 254-501-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f60a9a37053de0d86ebd11a56713644db762cca4&vid=f60a9a37053de0d86ebd11a56713644db762cca4&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 146.0 to 151.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR