Fmoc-D-Trp-OH ≥97.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: Nα-[(9H-芴-9-基甲氧基)羰基]-D-色氨酸, Fmoc-D-色氨酸

F0609-25G F0609-5G
TCIAF0609-25GEA 0 CNY
TCIAF0609-25G TCIAF0609-5G
Fmoc-D-Trp-OH ≥97.0% (by HPLC, titration analysis)
Fmoc-D-Trp-OH
分子式: C₂₆H₂₂N₂O₄
分子量: 426.46 g/mol
MDL编号: MFCD00062954
CAS号: 86123-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad6ac901b7206eceb28c14ec8f0245f325feabac&vid=ad6ac901b7206eceb28c14ec8f0245f325feabac&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 182.0 to 186.0 deg-C
Specific rotation [a]20/D +26.0 to +31.0 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR