4-(反-4-戊基环己基)苯腈 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-(trans-4-n-Pentylcyclohexyl)benzonitrile

A1828-25G A1828-5G
TCIAA1828-25GEA 0 CNY
TCIAA1828-25G TCIAA1828-5G
4-(反-4-戊基环己基)苯腈 ≥98.0% (by GC)
4-(反-4-戊基环己基)苯腈
分子式: C₁₈H₂₅N
分子量: 255.40 g/mol
溶点: 30°C
MDL编号: MFCD00075548
CAS号: 61204-01-1
EINECS: 262-656-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d77d1badec9b13c7dd1d8bd6abeb78f69a00b0c3&vid=d77d1badec9b13c7dd1d8bd6abeb78f69a00b0c3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Mesomorphic range 27.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR