Fmoc-Val-OH ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
Fmoc-L-缬氨酸, N-(9-芴甲氧羰基)-L-缬氨酸
F0299-25G
F0299-5G
TCIAF0299-25GEA
0
CNY
TCIAF0299-25G
TCIAF0299-5G
Fmoc-Val-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Val-OH
分子式:
C₂₀H₂₁NO₄ 分子量: 339.4 g/mol |
MDL编号:
MFCD00037124 CAS号: 68858-20-8 EINECS: 272-515-0 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 144.0 to 147.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...