Fmoc-D-Val-OH ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
Fmoc-D-缬氨酸, N-(9-芴甲氧羰基)-D-缬氨酸
F0610-25G
F0610-5G
TCIAF0610-25GEA
0
CNY
TCIAF0610-25G
TCIAF0610-5G
Fmoc-D-Val-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-D-Val-OH
分子式:
C₂₀H₂₁NO₄ 分子量: 339.39 g/mol |
MDL编号:
MFCD00062953 CAS号: 84624-17-9 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 144.0 to 148.0 deg-C |
Specific rotation [a]20/D | +15.0 to +19.0 deg (C=1, DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...