氰基联苯酚 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 4'-Hydroxy-4-biphenylcarbonitrile, 4'-Hydroxybiphenyl-4-carbonitrile, 4'-Hydroxy-[1,1'-biphenyl]-4-carbonitrile

H0523-25G H0523-5G
TCIAH0523-25GEA 0 CNY
TCIAH0523-25G TCIAH0523-5G
氰基联苯酚 ≥98.0% (by GC, titration analysis)
氰基联苯酚
分子式: C₁₃H₉NO
分子量: 195.22 g/mol
MDL编号: MFCD00059625
CAS号: 19812-93-2
EINECS: 427-840-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65168a3dea96701e636bb055bae860055997ce2e&vid=65168a3dea96701e636bb055bae860055997ce2e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 200.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR