1-乙酰基-4-哌啶甲酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: N-乙酰基-4-哌啶羧酸

A2261-25G A2261-5G
TCIAA2261-25GEA 0 CNY
TCIAA2261-25G TCIAA2261-5G
1-乙酰基-4-哌啶甲酸 ≥98.0% (by GC, titration analysis)
1-乙酰基-4-哌啶甲酸
分子式: C₈H₁₃NO₃
分子量: 171.19 g/mol
MDL编号: MFCD00023706
CAS号: 25503-90-6
EINECS: 247-049-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dac3b19aed342c0e234bb556ae546a4356cd45b&vid=5dac3b19aed342c0e234bb556ae546a4356cd45b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 182.0 to 186.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR