DL-乙硫氨基酪酸 ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: DL-2-氨基-4-(乙硫基)丁酸

E0038-25G E0038-5G
TCIAE0038-25GEA 0 CNY
TCIAE0038-25G TCIAE0038-5G
DL-乙硫氨基酪酸 ≥98.0% (by titrimetric analysis)
DL-乙硫氨基酪酸
分子式: C₆H₁₃NO₂S
分子量: 163.24 g/mol
MDL编号: MFCD00063102
CAS号: 67-21-0
EINECS: 200-647-0
Merck Index: 14,03738
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a3d1f315014179b4fc2415c13f470498bb6eeaa&vid=8a3d1f315014179b4fc2415c13f470498bb6eeaa&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR