N-Formyl-L-methionine ≥95.0% (by titrimetric analysis)

供应商: TCI

Synonyms: For-Met-OH, Formyl-L-methionine

F0124-1G F0124-5G
TCIAF0124-1GEA 0 CNY
TCIAF0124-1G TCIAF0124-5G
N-Formyl-L-methionine ≥95.0% (by titrimetric analysis)
N-Formyl-L-methionine
分子式: C₆H₁₁NO₃S
分子量: 177.2 g/mol
MDL编号: MFCD00021033
CAS号: 4289-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84dcd962f39bc9236b48c1552098a650ea0bcfed&vid=84dcd962f39bc9236b48c1552098a650ea0bcfed&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 95.0 %
Melting point 98.0 to 102.0 deg-C
Specific rotation [a]20/D -8.0 to -12.0 deg (C=0.8, H2O)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR