双(环戊二烯)二氯化铪(IV) ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 二氯二茂铪, 二(环戊二烯基)二氯化铪(IV)

H0914-1G H0914-25G H0914-5G
TCIAH0914-1GEA 0 CNY
TCIAH0914-1G TCIAH0914-25G TCIAH0914-5G
双(环戊二烯)二氯化铪(IV) ≥98.0% (by titrimetric analysis)
双(环戊二烯)二氯化铪(IV)
分子式: C₁₀H₁₀Cl₂Hf
分子量: 379.58 g/mol
MDL编号: MFCD00001438
CAS号: 12116-66-4
EINECS: 235-177-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a7f5d05b9470dbb78c466238d804f50278eedcb&vid=7a7f5d05b9470dbb78c466238d804f50278eedcb&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR