5-Bromo-2-fluoro-4-methylpyridine ≥98.0% (by GC)
供应商: TCI
|
Warning
|
B3579-1G
B3579-5G
TCIAB3579-1GEA
0
CNY
TCIAB3579-1G
TCIAB3579-5G
5-Bromo-2-fluoro-4-methylpyridine ≥98.0% (by GC)
5-Bromo-2-fluoro-4-methylpyridine
分子式:
C₆H₅BrFN 分子量: 190.01 g/mol |
MDL编号:
MFCD09839213 CAS号: 864830-16-0 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...